Learn More
Pepro Tech Inc Recombinant Murine FGF-10, 250ug DFS Item
Manufacturer:  Pepro Tech Inc 45061250UG
Recombinant Murine FGF-10 is a 17.0 kDa protein consisting of 148 amino acid residues.
Catalog No. 50-195-2586