Learn More
Pepro Tech Inc Recombinant Murine FGF-4, 250ug DFS Item
Manufacturer:  Pepro Tech Inc 45057250UG
Recombinant Murine FGF-4 is a 19.0 kDa protein consisting of 173 amino acid residues.
Catalog No. 50-195-2573