Learn More
Pepro Tech Inc Recombinant Murine IL-12 p80, 100ug DFS Item
Manufacturer:  Pepro Tech Inc 21012P80H100UG
Recombinant Murine IL-12 p80 is an 80.0 kDa, disulfide linked homodimer of two p40 chains of IL-12 that consists of 626 amino acid residues and less than 5% of the IL-12 p40 monomer.
Catalog No. 50-195-2480