Learn More
Pepro Tech Inc Recombinant Murine IL-17D, 50ug DFS Item
Manufacturer:  Pepro Tech Inc 21017D5UG
Recombinant Murine IL‐17D is a 20.2 kDa protein consisting of 184 amino acid residues.
Catalog No. 50-195-2492