Learn More
Pepro Tech Inc Recombinant Murine IL-17E, 25ug DFS Item
Manufacturer:  Pepro Tech Inc 21017E25UG
Recombinant Murine IL‐17E is a 35.0 kDa disulfide‐linked homodimer of two 152-amino-acid polypeptide chains.
Catalog No. 50-195-2496