Learn More
Novus Biologicals™ Recombinant Canine GM-CSF Animal-Free Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP27625420UG
 View more versions of this product
Catalog No. NB27625420U
Add to cart
Description
Description
An un-tagged bioactive recombinant protein corresponding to Canine GM-CSF
The Recombinant Canine GM-CSF Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
Lyoph
1437
>95%, by SDS-PAGE
E. coli
CSF2
Sterile water at 0.1 mg/mL
SDS-Page, Bioactivity
14.4 kDa
20 μg
Store at -20 to -80°C. Avoid freeze-thaw cycles.
GM-CSF is a monomer, 14.4 kDa (128 aa)
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5