Learn More
R&D Systems™ Recombinant Cynomolgus LAG-3 His-tag Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Applications: Bioactivity
Manufacturer:  R&D Systems™ 9992L3050
Catalog No. 99-92L-3050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Cynomolgus LAG-3 His-tag Protein is derived from CHO. The Recombinant Cynomolgus LAG-3 His-tag Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
XP_005570011.1
47 kDa
50 μg
12 months from date of receipt, <= -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
<0.10 EU per 1 μg of the protein by the LAL method.
LAG-3
Cynomolgus Monkey
Reconstitute at 500 μg/mL in PBS.
3902
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
CHO
RUO
Bioactivity
Unconjugated
Yes
Lyophilized from a 0.2 μm filtered solution in PBS.