Learn More
R&D Systems™ Recombinant Cynomolgus Siglec-2/CD22 His-tag Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9864SL050
Catalog No. 98-64S-L050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Cynomolgus Monkey Siglec-2/CD22 Protein is derived from HEK293. The Recombinant Cynomolgus Monkey Siglec-2/CD22 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
EHH59463
Lyophilized from a 0.2μm filtered solution in PBS.
76 kDa
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
HEK293
<0.10 EU per 1μg of the protein by the LAL method.
Cynomolgus Monkey
Bioactivity
933
B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, Siglec2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Reconstitute at 500μg/mL in PBS.