Learn More
Novus Biologicals™ Recombinant Dopa Decarboxylase/DDC Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
Manufacturer:  Novus Biologicals™ NBP244296
Catalog No. NBP244296
Add to cart
Description
Description
A recombinant protein with a His-tag corresponding to amino acids 1 to 480 of Human Dopa Decarboxylase/DDC The Recombinant Human Dopa Decarboxylase/DDC Protein is derived from E. coli. The Recombinant Human Dopa Decarboxylase/DDC Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
1mg/mL
1644
0.5mg
RUO - research use only
20mM Tris-HCl buffer (pH 8.0) containing 2mM DTT, 10% glycerol
>95% pure by SDS-PAGE
Store at 4° short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Yes