Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant E. coli mutM Protein

Highly purified. Generating reliable and reproducible results.

$356.50 - $1004.00

Specifications

Source E.coli
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol mutM
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25951250 View Documents Novus Biologicals™
NBP25951250UG
50μg Each for $356.50
Add to cart
 
NBP259512Y View Documents Novus Biologicals™
NBP259512250UG
250μg Each for $1,004.00
Add to cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-269 of E.coli mutM The Recombinant E. coli mutM Protein is derived from E. coli. The Recombinant E. coli mutM Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

E.coli
RUO
mutM
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
32.4 kDa
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
946765
Recombinant E. coli mutM Protein
Liquid. 20mM Tris-HCl buffer (pH8.0) containing 20% glycerol, 0.1M NaCl,1mM DTT
Unlabeled
0.5mg/mL

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit