Learn More
Active Motif Recombinant FXR protein
SDP
Supplier:  Active Motif 31120
Encompass_Preferred
Recombinant FXR is isolated from an E. coli strain that carries the coding sequence of the human FXR under the control of a T7 promoter (accession number U68233). The purified recombinant protein has an amino terminal polyhistidine tag and is greater than 90% homogeneous and contains no detectable protease, DNase and RNase activity. The farnesoid X receptor (FXR) or nuclear receptor subfamily 1, group H, member 4 (NR1H4) is a nuclear receptor that, when activated, translocates to the nucleus where it interacts with certain proteins, including RXR and PPARGC1A, forming heterodimers that bind to hormone response elements on DNA and modulate gene expression. One of the primary functions of FXR is to suppress bile acid synthesis when levels are high by inhibiting the transcription of cholesterol 7 alpha-hydroxylase (CYP7A1) gene that expresses CYP7A1, the rate-limiting enzyme in bile acid synthesis from cholesterol.
Catalog No. NC1842957
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.