Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human 2B4/CD244/SLAMF4 Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1025.00

Specifications

Source Baculovirus
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol CD244
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25957350 View Documents Novus Biologicals™
NBP25957350UG
50μg Each for $287.00
Add to cart
 
NBP259573Y View Documents Novus Biologicals™
NBP259573250UG
250μg Each for $1,025.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 19-224 of Human 2B4/CD244/SLAMF4 The Recombinant Human 2B4/CD244/SLAMF4 Protein is derived from Baculovirus. The Recombinant Human 2B4/CD244/SLAMF4 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Baculovirus
RUO
CD244
51744
Recombinant Human 2B4/CD244/SLAMF4 Protein
Liquid. PBS (pH 7.4) containing 10% glycerol
Unlabeled
1mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activ
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
50.2 kDa
<1.0 EU per 1μg of protein (determined by LAL method)

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit