Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Recombinant Human Adiponectin/Acrp30 Animal-Free Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
$83.90 - $3163.70
Specifications
Buffer | Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, 0.75 mM DTT, pH 8.0 |
---|---|
Concentration | Lyoph |
For Use With (Application) | SDS-Page, Bioactivity |
Gene ID (Entrez) | 9370 |
Gene Symbol | ADIPOQ |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2762105U
|
Novus Biologicals
NBP2762105UG |
5 μg |
Each for $83.90
|
|
NB27621025U
|
Novus Biologicals
NBP27621025UG |
25 μg |
Each for $190.10
|
|
NBP2762101M
|
Novus Biologicals
NBP2762101MG |
1 mg |
Each for $3,163.70
|
|
Description
An un-tagged bioactive recombinant protein corresponding to Human Adiponectin/Acrp30The Recombinant Human Adiponectin/Acrp30 Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, 0.75 mM DTT, pH 8.0 | |
SDS-Page, Bioactivity | |
ADIPOQ | |
>95%, by SDS-PAGE | |
E. coli | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. |
Lyoph | |
9370 | |
16.7 kDa | |
Sterile 5 mM Tris and 0.75 mM DTT, pH 8 at 0.1 mg/mL | |
Adiponectin/Acrp30 is a monomer, 16.7 kDa (145 aa) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title