Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human alpha-Synuclein Aggregate Protein (Biologically Active)

Highly purified and high bioactivity. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP2547892X100UGVIALS

 View more versions of this product

Catalog No. NB254789210

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

An un-tagged full length Human biologically active Alpha-Synuclein recombinant protein aggregate (pre-formed fibrils, Type 1), NCBI Accession #: NP_000336.1. The Recombinant Human alpha-Synuclein Aggregate Protein is derived from E. coli. The Recombinant Human alpha-Synuclein Aggregate Protein has been validated for the following applications: Western Blot, In vitro assay, In vivo assay, SDS-Page, Bioactivity.
Specifications

Specifications

Western Blot, In vitro assay, In vivo assay, SDS-Page
6622
>95% pure by SDS-PAGE
E.Coli
Store at −80°C. Avoid freeze-thaw cycles.
alpha-Synuclein, Lewy body 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor)
Endogenous alpha-synuclein phosphorylation. 100μM alpha synuclein protein monomer seeded with 10nM alpha synuclein protein aggregate in 25μM Thioflavin T (PBS pH 7.4, 100μL reaction volume) generated a fluorescence intensity of 13,000 Relative Fluorescence Units after incubation at 37°C with shaking at 600 rpm for 24 hours. Fluorescence was measured by excitation at 450nm and emission at 485nm on a Molecular Devices Gemini XPS microplate reader.
Unlabeled
Yes
PBS
Human alpha-Synuclein Aggregate Protein
2 x 100μg Vials
MDVFMKGLSKAKEGVVAAAEKTKQGVAEAAGKTKEGVLYVGSKTKEGVVHGVATVAEKTKEQVTNVGGAVVTGVTAVAQKTVEGAGSIAAATGFVKKDQLGKNEEGAPQEGILEDMPVDPDNEAYEMPSEEGYQDYEPEA
RUO
SNCA
Recombinant Protein
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit