Learn More
R&D Systems™ Recombinant Human alpha-Synuclein Low Endotoxin Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Applications: Bioactivity
Manufacturer:  R&D Systems™ SP486500
Catalog No. SP486500
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human alpha-Synuclein Low Endotoxin Protein is derived from E. coli. The Recombinant Human alpha-Synuclein Low Endotoxin Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
P37840
14 kDa
500 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 24 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening.
Bioactivity
Unconjugated
Yes
6622
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain.
E. coli
RUO
alpha-Synuclein
Human
X mg/mL (X μm) in 50 mM HEPES pH 7.5, 100 mM NaCl, 10% (v/v) Glycerol, 2 mM TCEP