Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Recombinant Human AlphaB Crystallin/CRYAB His Protein
$79.90 - $595.00
Specifications
Concentration | 1 mg/ml |
---|---|
For Use With (Application) | SDS-PAGE |
Formulation | PBS (pH 7.4), 10% glycerol |
Gene ID (Entrez) | 1410 |
Molecular Weight (g/mol) | 21.2 kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB355227
|
Novus Biologicals
NBP32459950UG |
50 μg |
Each for $211.50
|
|
|||||
NB355228
|
Novus Biologicals
NBP324599250UG |
250 μg |
Each for $595.00
|
|
|||||
NB355229
|
Novus Biologicals
NBP32459920UG |
20 μg |
Each for $150.50
|
|
|||||
NB355230
|
Novus Biologicals
NBP32459910UG |
10 μg |
Each for $79.90
|
|
|||||
NB355231
|
Novus Biologicals
NBP324599100UG |
100 μg |
Each for $342.50
|
|
|||||
Specifications
1 mg/ml | |
PBS (pH 7.4), 10% glycerol | |
21.2 kDa | |
CRYAB | |
>90%, by SDS-PAGE |
SDS-PAGE | |
1410 | |
alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component | |
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision | |
AlphaB Crystallin/CRYAB |
Spot an opportunity for improvement?
Product Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title