Learn More
Novus Biologicals™ Recombinant Human Amphiregulin Animal-Free Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP27626450UG
 View more versions of this product
Catalog No. NB27626450U
Add to cart
Description
Description
An un-tagged bioactive recombinant protein corresponding to Human Amphiregulin
The Recombinant Human Amphiregulin Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
Lyoph
374
>95%, by SDS-PAGE
E. coli
AREG
Sterile water at 0.1 mg/mL
SDS-Page, Bioactivity
10.1 kDa
50 μg
Store at -20 to -80°C. Avoid freeze-thaw cycles.
Amphiregulin is a monomer, 10.1 kDa (87 aa)
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5