Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Artemin Protein

Highly purified and high bioactivity. Generating reliable and reproducible results.

$83.00 - $2970.65

Specifications

Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium citrate, 25 mM sodium chloride, pH 4.5
Concentration Lyoph
For Use With (Application) SDS-Page
Gene ID (Entrez) 9048
Gene Symbol ARTN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP2762565U
View Documents
Novus Biologicals™
NBP2762565UG
5 μg
Each for $83.00
Add to cart
 
NB27625620U
View Documents
Novus Biologicals™
NBP27625620UG
20 μg
Each for $188.00
Add to cart
 
NBP2762561M
View Documents
Novus Biologicals™
NBP2762561MG
1 mg
Each for $2,970.65
Add to cart
 
Description

Description

An un-tagged recombinant protein corresponding to Human Artemin

The Recombinant Human Artemin Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium citrate, 25 mM sodium chloride, pH 4.5
SDS-Page
ARTN
>95%, by SDS-PAGE
E. coli
Store at -20 to -80°C. Avoid freeze-thaw cycles.
Lyoph
9048
24.2 kDa
Sterile water at 0.1 mg/mL
Artemin is a dimer, 12.1/24.2 kDa (113/226 aa)
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit