Learn More
R&D Systems™ Recombinant Human B7-H6 His-tag Protein CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9309B701M
Catalog No. 93-09B-701M
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human B7-H6 His-tag Protein is derived from HEK293. The Recombinant Human B7-H6 His-tag Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Q68D85
Bioactivity
374383
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Human embryonic kidney cell, HEK293-derived Asp25-Ser262, with a C-terminal 6-His tag
<0.10EU per 1μg of the protein by the LAL method.
Recombinant Protein
Human
Bioactivity
Lyophilized from a 0.2μm filtered solution in PBS.
28kDa
1mg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1
Unconjugated
Reconstitute at 200μg/mL in PBS.