Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human beta-1,3-Glucuronyltransferase 1/B3GAT1 Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$366.50 - $1784.00

Specifications

Concentration 0.25 mg/mL
Endotoxin Level < 1.0 EU per 1 μg of protein (determined by LAL method)
Purity >90% SDS-PAGE
Source Baculovirus
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB002931
View Documents
Novus Biologicals™
NBP2774890.05MG
0.05 mg
Each for $366.50
Add to cart
 
NB002932
View Documents
Novus Biologicals™
NBP2774890.1MG
0.1 mg
Each for $576.50
Add to cart
 
NB002933
View Documents
Novus Biologicals™
NBP2774890.5MG
0.5 mg
Each for $1,784.00
Add to cart
 
Description

Description

A recombinant protein with a hIgG-His-Tag and corresponding to the amino acids 28-334aa of Human B3GAT1.

Source: Baculovirus

Amino Acid Sequence:

ADP>TLAPLLA VHKDEGSDPR RETPPGADPR EYCTSDRDIV EVVRTEYVYT RPPPWSDTLP TIHVVTPTYS RPVQKAELTR MANTLLHVPN LHWLVVEDAP RRTPLTARLL RDTGLNYTHL HVETPRNYKL RGDARDPRIP RGTMQRNLAL RWLRETFPRN SSQPGVVYFA DDDNTYSLEL FEEMRSTRRV SVWPVAFVGG LRYEAPRVNG AGKVVGWKTV FDPHRPFAID MAGFAVNLRL ILQRSQAYFK LRGVKGGYQE SSLLRELVTL NDLEPKAANC TKILVWHTRT EKPVLVNEGK KGFTDPSVEI LEPKSCDKTH TCPPCPAPEL LGGPSVFLFP PKPKDTLMIS RTPEVTCVVVDVSHEDPEVK FNWYVDGVEV HNAKTKPREE QYNSTYRVVS VLTVLHQDWL NGKEYKCKVSNKALPAPIEK TISKAKGQPR EPQVYTLPPS RDELTKNQVS LTCLVKGFYP SDIAVEWESNGQPENNYKTT PPVLDSDGSF FLYSKLTVDK SRWQQGNVFS CSVMHEALHNHYTQKSLSLS PGKHHHHHH>

Specifications

Specifications

0.25 mg/mL
< 1.0 EU per 1 μg of protein (determined by LAL method)
>90% SDS-PAGE
Baculovirus
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit