Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human beta-NGF Animal-Free Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBP27624220UG
Description
An un-tagged bioactive recombinant protein corresponding to Human beta-NGFThe Recombinant Human beta-NGF Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Lyoph | |
4803 | |
>95%, by SDS-PAGE | |
E. coli | |
NGF | |
Sterile water at 0.1 mg/mL |
Bioactivity, SDS-PAGE | |
27.3 kDa | |
20 μg | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
beta-NGF is a dimer, 13.6/27.3 kDa (121/242 aa) | |
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction