Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BMP-4 (E. coli-expressed) Protein, CF

Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells.

$219.50 - $513.50

Specifications

Accession Number P12644
Buffer Lyophilized from a 0.2 µm filtered solution in Acetonitrile and TFA.
Conjugate Unconjugated
Endotoxin Level <0.10 EU per 1 µg of the protein by the LAL method.
For Use With (Application) Bioactivity
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
31-4BP-E010
View Documents
R&D Systems™
314BPE010
10 µg
Each for $219.50
Add to cart
 
31-4BP-E050
View Documents
R&D Systems™
314BPE050
50 µg
Each for $513.50
Add to cart
 
Description

Description

Specifications

Specifications

P12644
Unconjugated
Bioactivity
12 kDa
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
E. coli-derived human BMP-4 protein Lys303-Arg408
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -70 °C under sterile conditions after reconstitution.
Lyophilized from a 0.2 µm filtered solution in Acetonitrile and TFA.
<0.10 EU per 1 µg of the protein by the LAL method.
652
BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
Reconstitute at 100 µg/mL in 4 mM HCl.
Human
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit