Learn More
Pepro Tech Inc Recombinant Human BMP-7, 250ug DFS Item
Manufacturer:  Pepro Tech Inc 12003250UG
Recombinant Human BMP-7 is a 28.8 kDa homodimeric glycoprotein consisting of two 117 amino acid subunits, which correspond to amino acid residues 315 to 431 of the full-length BMP-7 precursor.
Catalog No. 50-195-2472