Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human BMPR-IA/ALK-3 Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1025.00

Specifications

Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol BMPR1A
Gene Alias ACVRLK310q23del, ALK-3, ALK3EC 2.7.11.30, BMP type-1A receptor, BMPR-1A, bone morphogenetic protein receptor type-1A, bone morphogenetic protein receptor, type IA, CD292, CD292 antigen, EC 2.7.11, Serine/threonine-protein kinase receptor R5, SKR5, type II
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25956250 View Documents Novus Biologicals™
NBP25956250UG
50μg Each for $287.00
Add to cart
 
NBP259562Y View Documents Novus Biologicals™
NBP259562250UG
250μg Each for $1,025.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids24-152 of Human BMPR-IA/ALK-3 The Recombinant Human BMPR-IA/ALK-3 Protein is derived from Baculovirus. The Recombinant Human BMPR-IA/ALK-3 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
ACVRLK310q23del, ALK-3, ALK3EC 2.7.11.30, BMP type-1A receptor, BMPR-1A, bone morphogenetic protein receptor type-1A, bone morphogenetic protein receptor, type IA, CD292, CD292 antigen, EC 2.7.11, Serine/threonine-protein kinase receptor R5, SKR5, type II
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
41.4 kDa
<1.0 EU per 1μg of protein (determined by LAL method)
RUO
BMPR1A
657
Recombinant Human BMPR-IA/ALK-3 Protein
Liquid. PBS (pH 7.4) containing 10% glycerol
Unlabeled
1mg/mL
Human

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit