Learn More
Pepro Tech Inc Recombinant Human C5a, 500ug DFS Item
Manufacturer:  Pepro Tech Inc 30070500UG
Recombinant Human C5a is an 8.3 kDa glycoprotein containing the 74 amino acid residues of the C5a anaphylatoxin chain.
Catalog No. 50-195-2522