Learn More
R&D Systems™ Recombinant Human CD155/PVR Fc Chimera Protein CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Manufacturer:  R&D Systems™ 9174CD01M
Catalog No. 91-74C-D01M
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human CD155/PVR Fc Chimera Protein is derived from HEK293. The Recombinant Human CD155/PVR Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Bioactivity
Lyophilized from a 0.2μm filtered solution in PBS.
61kDa
1mg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVR, PVS, PVSFLJ25946, Tage4
Unconjugated
Reconstitute at 500μg/mL in PBS.
Bioactivity
5817
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
HEK293
<0.10EU per 1μg of the protein by the LAL method.
Recombinant Protein
Human