Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human CD40 Ligand/TNFSF5 (Histidine-tagged), CF
A New rh CD40 Ligand is Now Available! ∼10 fold better activity; HEK293 expressed; HA tag!
Supplier: R&D Systems™ 2706CL500/CF
This item is not returnable.
View return policy
Description
Measured in a cell proliferation assay using enriched human B cells in the presence of IL-4. Spriggs, M.K. et al. (1992) J. Exp. Med. 176:1543. The ED50 for this effect is 2-12 ng/mL in the presence of 10 μg/mL of a cross-linking antibody, Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ) and 20 ng/mL of recombinant human IL-4.Specifications
P29965 | |
Lyophilized from a 0.2 µm filtered solution in HEPES, NaCl and EDTA. | |
M.W. (Observed): 18 kDa, reducing conditions; M.W. (theoretical): 17.7 kDa (monomer) | |
>97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. | |
E. coli-derived human CD40 Ligand/TNFSF5 protein Glu108-Leu261, with an N-terminal Met and 6-His tag | |
<0.01 EU / 1 µg of the protein by the LAL method. | |
Unconjugated | |
Reconstitute at 100 µg/mL in sterile PBS. |
Bioactivity | |
959 | |
CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40L, CD40-L, CD40LG, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5, TNFSF5IMD3, TRAP, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5 | |
500 µg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. | |
CD40LG | |
Human | |
CD40 Ligand/TNFSF5 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title