Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CD84/SLAMF5 Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$358.00 - $1383.00

Specifications

Source Baculovirus
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol CD84
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25956950 View Documents Novus Biologicals™
NBP25956950UG
50μg Each for $358.00
Add to cart
 
NBP259569Y View Documents Novus Biologicals™
NBP259569250UG
250μg Each for $1,383.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 22-225 of Human CD84/SLAMF5 The Recombinant Human CD84/SLAMF5 Protein is derived from Baculovirus. The Recombinant Human CD84/SLAMF5 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Baculovirus
RUO
CD84
8832
Recombinant Human CD84/SLAMF5 Protein
Liquid. PBS (pH 7.4) containing 10% glycerol
Unlabeled
0.5mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocyti
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
23.8 kDa
<1.0 EU per 1μg of protein (determined by LAL method)

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit