Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Novus Biologicals™ Recombinant Human CEACAM-5/CD66e Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1025.00

Specifications

Source Baculovirus
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol CEACAM5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25956750 View Documents Novus Biologicals™
NBP25956750UG
50μg Each for $287.00
N/A
NBP259567Y View Documents Novus Biologicals™
NBP259567250UG
250μg Each for $1,025.00
N/A
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 35-685 of Human CEACAM5/CD66e The Recombinant Human CEACAM5/CD66e Protein is derived from Baculovirus. The Recombinant Human CEACAM5/CD66e Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Baculovirus
RUO
CEACAM5
1048
Recombinant Human CEACAM-5/CD66e Protein
Liquid. PBS (pH 7.4) containing 10% glycerol
Unlabeled
0.5mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
72.3 kDa
<1.0 EU per 1μg of protein (determined by LAL method)

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit