Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Chemerin Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1025.00

Specifications

Source Baculovirus
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol RARRES2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25954720 View Documents Novus Biologicals™
NBP25954720UG
20μg Each for $287.00
Add to cart
 
NBP259547X View Documents Novus Biologicals™
NBP259547100UG
100μg Each for $1,025.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 21-157 of Human Chemerin The Recombinant Human Chemerin Protein is derived from E. coli. The Recombinant Human Chemerin Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Baculovirus
RUO
RARRES2
5919
Recombinant Human Chemerin Protein
Liquid. PBS (pH 7.4) containing 10% glycerol
Unlabeled
0.5mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
chemerin, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG2HP10433
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
16.9 kDa
< 1.0 EU per 1 microgram of protein (determined by LAL method)

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit