Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Chorionic Gonadotropin beta Chain (HCG beta) Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$285.00 - $1004.00

Specifications

Source Baculovirus
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol CGB
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25954550 View Documents Novus Biologicals™
NBP25954550UG
50μg Each for $1,004.00
Add to cart
 
NBP25954510 View Documents Novus Biologicals™
NBP25954510UG
10μg Each for $285.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 21-165 of Human Chorionic Gonadotropin beta Chain (HCG beta) The Recombinant Human Chorionic Gonadotropin beta Chain (hCG beta) Protein is derived from E. coli. The Recombinant Human Chorionic Gonadotropin beta Chain (hCG beta) Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Baculovirus
RUO
CGB
1082
Recombinant Human Chorionic Gonadotropin beta Chain (HCG beta) Protein
Liquid. PBS (pH 7.4) containing 10% glycerol
Unlabeled
0.25mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
CGB3choriogonadotropin subunit beta, CGB5, CGB7, CGB8, CG-beta, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 3 subunit, chorionic gonadotropin beta chain, chorionic gonadotropin beta subunit, chorionic gonadotropin, beta polypeptide, hC
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
16.6 kDa
< 1.0 EU per 1 microgram of protein (determined by LAL method)

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit