Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Cleavage Stimulation Factor Protein
SDP

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$494.50 - $1932.00

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. PBS (pH 7.4) containing 1mM DTT, 20% glycerol
Gene ID (Entrez) 1477
Molecular Weight (g/mol) 49.1 kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25959450
SDP
View Documents
Novus Biologicals™
NBP25959450UG
50μg
Each for $494.50
Add to Cart
 
NBP259594Y
SDP
View Documents
Novus Biologicals™
NBP259594250UG
250μg
Each for $1,932.00
Add to Cart
 
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-431 of Human cleavage stimulation factor The Recombinant Human cleavage stimulation factor Protein is derived from E. coli. The Recombinant Human cleavage stimulation factor Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.5mg/mL
Liquid. PBS (pH 7.4) containing 1mM DTT, 20% glycerol
49.1 kDa
>90%, by SDS-PAGE
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
<1.0 EU per 1μg of protein (determined by LAL method)
CSTF1
Unconjugated
Recombinant
SDS-PAGE
1477
Recombinant Human cleavage stimulation factor Protein
Baculovirus
RUO
CF-1 50 kDa subunit, Cleavage stimulation factor 50 kDa subunit, cleavage stimulation factor subunit 1, cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kD, cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa, CSTF 50 kDa subunit, CstF-50, CstF
Recombinant Protein
Human
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit