Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human DCXR Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

$129.70 - $351.00

Specifications

Source E.coli
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol DCXR
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25954320 View Documents Novus Biologicals™
NBP25954320UG
20μg Each for $129.70
Add to cart
 
NBP259543X View Documents Novus Biologicals™
NBP259543100UG
100μg Each for $351.00
Add to cart
 
Description

Description

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-244 of Human DCXR The Recombinant Human DCXR Protein is derived from E. coli. The Recombinant Human DCXR Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

E.coli
RUO
DCXR
51181
Recombinant Human DCXR Protein
Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 20% glycerol, 50mM NaCl
Unlabeled
0.5mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Carbonyl reductase II, DCR, dicarbonyl/L-xylulose reductaseP34H, EC 1.1.1, EC 1.1.1.10, HCR2, HCRII, human carbonyl reductase 2, KIDCR, Kidney dicarbonyl reductase, L-xylulose reductase, SDR20C1, short chain dehydrogenase/reductase family 20C, member 1, S
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
28 kDa

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit