Learn More
Novus Biologicals Defensin alpha 3 Synthetic Peptide
Human Recombinant Protein
Manufacturer: Novus Biologicals NBP262784
Description
Chain A of DEFA3. Sequence: CYCRIPACIAGERRYGTCIYQGRLWAFCC The Defensin alpha 3 Synthetic Peptide is derived from Synthetic.Specifications
LYOPH | |
50 μg | |
Store the unopened product at -20°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Storage below -20°C is not recommended. Do not use past expiration date. | |
DEFA3 | |
Human |
1668 | |
Synthetic | |
RUO | |
Defensin alpha 3 | |
Reconstitute the peptide with distilled or de-ionized water or small percentage organic solvent (eg DMSO) dependent on the intended application of the peptide (Caution: vial is under vacuum). |