Learn More
R&D Systems™ Recombinant Human DNAM-1/CD226 His-tag Protein, Carrier-free Available on GSA/VA Contract for Federal Government customers only.
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9298DN050
Catalog No. 92-98D-N050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human DNAM-1/CD226 His-tag Protein is derived from HEK293. The Recombinant Human DNAM-1/CD226 His-tag Protein has been validated for the following applications: Bioactivity.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Ships at ambient temperature
Specifications
Specifications
Q15762
Lyophilized from a 0.2μm filtered solution in PBS
27 kDa
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Human embryonic kidney cell, HEK293-derived Glu19-Asn247, with C-terminal 6-His tag
RUO
CD226 antigen platelet and T cell activation antigen 1; CD226 molecule; CD226; DNAM1; DNAM-1; DNAM1adhesion glycoprotein; DNAM-1DNAX accessory molecule-1; DNAX accessory molecule 1; PTA1; T lineage-specific activation antigen 1 antigen; TLiSA1 Related Res
Recombinant Protein
Human
Bioactivity
10666
DNAM-1/CD226 His-tag Recombinant Protein
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
<0.10 EU per 1μg of the protein by the LAL method
0.4 to 2μg/mL
Unconjugated
Yes
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.