Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human DUSP18 Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

$129.70 - $351.00

Specifications

Source E.coli
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol DUSP18
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25958320 View Documents Novus Biologicals™
NBP25958320UG
20μg Each for $129.70
Add to cart
 
NBP259583X View Documents Novus Biologicals™
NBP259583100UG
100μg Each for $351.00
Add to cart
 
Description

Description

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-188 of Human DUSP18 The Recombinant Human DUSP18 Protein is derived from E. coli. The Recombinant Human DUSP18 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

E.coli
RUO
DUSP18
150290
Recombinant Human DUSP18 Protein
Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 40% glycerol, 0.1mM PMSF, 1mM EDTA
Unlabeled
0.5mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
bK963H5.1, DSP18, dual specificity phosphatase 18, dual specificity protein phosphatase 18, DUSP20, EC 3.1.3.16, EC 3.1.3.48, LMWDSP20, LMW-DSP20, Low molecular weight dual specificity phosphatase 20, MGC32658
Recombinant Protein
>90%, by SDS-PAGE
Functional, SDS-Page
23.6 kDa
Human

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit