Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human EBI3 HIS-tagged Protein

Highly purified and high bioactivity. Generating reliable and reproducible results.

$83.00 - $240.50

Specifications

Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, 30 mM sodium chloride, pH7.5
Concentration Lyoph
For Use With (Application) Western Blot, SDS-Page
Gene ID (Entrez) 10148
Gene Symbol EBI3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP2762312U
View Documents
Novus Biologicals™
NBP2762312UG
2 μg
Each for $83.00
Add to cart
 
NB27623110U
View Documents
Novus Biologicals™
NBP27623110UG
10 μg
Each for $240.50
Add to cart
 
Description

Description

A HIS-tagged recombinant protein corresponding to Human IL-35

The Recombinant Human EBI3 Protein has been validated for the following applications: Western Blot, SDS-Page.
Specifications

Specifications

Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, 30 mM sodium chloride, pH7.5
Western Blot, SDS-Page
EBI3
>95%, by SDS-PAGE
HEK293
Store at -20 to -80°C. Avoid freeze-thaw cycles.
Lyoph
10148
45.8 kDa
Sterile water at 0.1 mg/mL
IL-35 is a single chain dimer, Predicted MW of 49 kDa and an observed MW of ∼ 75-95 kDa reduced or ∼ 65-75 kDa non-reduced, depending on type of gel and analysis. (406 aa)
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit