Learn More
Novus Biologicals™ Recombinant Human ECHS1 Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
$179.60 - $458.00
Specifications
Concentration | 1mg/mL |
---|---|
For Use With (Application) | Functional Study, SDS-Page |
Formulation | Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 20% glycerol, 100mM NaCl |
Gene ID (Entrez) | 1892 |
Molecular Weight (g/mol) | 30.6 kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25958820
|
Novus Biologicals™
NBP25958820UG |
20μg |
Each for $179.60
|
|
|||||
NBP259588X
|
Novus Biologicals™
NBP259588100UG |
100μg |
Each for $458.00
|
|
|||||
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 28-290 of Human ECHS1 The Recombinant Human ECHS1 Protein is derived from E. coli. The Recombinant Human ECHS1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.Specifications
1mg/mL | |
Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 20% glycerol, 100mM NaCl | |
30.6 kDa | |
>90%, by SDS-PAGE | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
EC 4.2.1, enoyl CoA hydratase, short chain, 1, mitochondrial, enoyl Coenzyme A hydratase, short chain, 1, mitochondrial, Enoyl-CoA hydratase 1, enoyl-CoA hydratase, mitochondrial, SCEHEC 4.2.1.17, Short-chain enoyl-CoA hydratase | |
Recombinant Protein | |
Human |
Functional Study, SDS-Page | |
1892 | |
Recombinant Human ECHS1 Protein | |
E.coli | |
RUO | |
ECHS1 | |
Unconjugated | |
Recombinant |
For Research Use Only.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.