Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human EDA2R/TNFRSF27/XEDAR Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1025.00

Specifications

Source Baculovirus
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol EDA2R
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25955050 View Documents Novus Biologicals™
NBP25955050UG
50μg Each for $287.00
Add to cart
 
NBP259550Y View Documents Novus Biologicals™
NBP259550250UG
250μg Each for $1,025.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 1-138 of Human EDA2R/TNFRSF27/XEDAR The Recombinant Human EDA2R/TNFRSF27/XEDAR Protein is derived from Baculovirus. The Recombinant Human EDA2R/TNFRSF27/XEDAR Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Baculovirus
RUO
EDA2R
60401
Recombinant Human EDA2R/TNFRSF27/XEDAR Protein
Liquid. PBS (pH 7.4) containing 20% glycerol, 1mM DTT.
Unlabeled
0.5mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
ectodysplasin A2 receptor, EDA-A2 receptor, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
42.5 kDa
< 1.0 EU per 1 microgram of protein (determined by LAL method)

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit