Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human FABP4/A-FABP Protein

Highly purified and high bioactivity. Generating reliable and reproducible results.

Applications: SDS-Page

Manufacturer:  Novus Biologicals™ NBP199612100UG

Catalog No. N199612100U


Add to cart

Description

Description

Recombinant protein for Human FABP4/A-FABP

The Recombinant Human FABP4/A-FABP Protein is derived from E. coli. The Recombinant Human FABP4/A-FABP Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

SDS-Page
2167
>98%, by SDS-PAGE
E. coli
RUO
FABP4
Unconjugated
Yes
Sterile filtered and lyophilized with no additives.
Lyoph
16 kDa
100 μg
Store at -20 to -80°C. Avoid freeze-thaw cycles.
<0.1 ng/ug
FABP4/A-FABP
Human
Reconstitute in dH2O to a concentration of 0.1 - 1.0 mg/mL. The solution can then be diluted into other aqueous buffers and stored at 4C for 1 week or -20°C for long-term storage.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit