Learn More
Novus Biologicals™ Recombinant Human FGF-5 Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP2762771MG
 View more versions of this product
Catalog No. NBP2762771M
Add to cart
Description
Description
An un-tagged bioactive recombinant protein corresponding to Human FGF-5
The Recombinant Human FGF-5 Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
Lyoph
2250
>95%, by SDS-PAGE
E. coli
FGF5
Sterile water at 0.1 mg/mL. If a precipitate is observed, centrifuge the solution thoroughly and use only the soluble fraction (removing it from the precipitate). A 10% overfill has been added to compensate for any loss of protein in the precipitate.
SDS-Page, Bioactivity
27.7 kDa
1 mg
Store at -20 to -80°C. Avoid freeze-thaw cycles.
FGF-5 is a monomer, 27.7 kDa (252 aa)
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate and 100 mM sodium chloride, pH 7.5