Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human FGF-8 Animal-Free Protein

Highly purified and high bioactivity. Generating reliable and reproducible results.

$83.00 - $3242.79

Specifications

Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 5 mM sodium phosphate, 50mM soldium chloride, pH 7.5
Concentration Lyoph
For Use With (Application) SDS-Page, Bioactivity
Gene ID (Entrez) 2253
Gene Symbol FGF8
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP2762495U
View Documents
Novus Biologicals™
NBP2762495UG
5 μg
Each for $83.00
Add to cart
 
NB27624925U
View Documents
Novus Biologicals™
NBP27624925UG
25 μg
Each for $188.00
Add to cart
 
NBP2762491M
View Documents
Novus Biologicals™
NBP2762491MG
1 mg
Each for $3,242.79
Add to cart
 
Description

Description

An un-tagged bioactive recombinant protein corresponding to Human FGF-8

The Recombinant Human FGF-8 Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications

Specifications

Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 5 mM sodium phosphate, 50mM soldium chloride, pH 7.5
SDS-Page, Bioactivity
FGF8
>95%, by SDS-PAGE
E. coli
Store at -20 to -80°C. Avoid freeze-thaw cycles.
Lyoph
2253
22.5 kDa
Sterile water at 0.1 mg/mL. If a precipitate is observed, centrifuge the solution thoroughly and use only the soluble fraction (removing it from the precipitate). A 10% overfill has been added to compensate for any loss of protein in the precipitate.
FGF-8 is a monomer, 22.5 kDa (194 aa)
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit