Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human FGF basic/FGF2/bFGF (145 aa) Protein
Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells.
Supplier: R&D Systems™ 3718FB100/DUP
This item is not returnable.
View return policy
Description
Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 0.1-0.6 ng/mL.Specifications
NP_001997 | |
Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl. | |
M.W. (theoretical): 16 kDa | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
FGF2 | |
Human | |
Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4, EDTA and DTT. | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Bioactivity | |
2247 | |
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin | |
E. coli-derived human FGF basic/FGF2/bFGF protein Ala144-Ser288 | |
<0.01 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
Reconstitute at 100 μg/mL in sterile PBS. | |
<0.01 EU per 1 μg of the protein by the LAL method. | |
FGF basic/FGF2/bFGF |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction