Learn More
R&D Systems™ Recombinant Human GFR alpha-like His-tag Protein CF Available on GSA/VA Contract for Federal Government customers only.
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9647GR050
Catalog No. 96-47G-R050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human GFR alpha-like His-tag Protein is derived from HEK293. The Recombinant Human GFR alpha-like His-tag Protein has been validated for the following applications: Bioactivity.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Q6UXV0
Lyophilized from a 0.2μm filtered solution in PBS.
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
HEK293
<0.10 EU per 1μg of the protein by the LAL method.
Human
Bioactivity
39 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Reconstitute at 500μg/mL in PBS.