Learn More
Novus Biologicals™ Recombinant Human GILT/IFI30 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP2231460.25MG
Catalog No. NB223146025
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 58-232 of Human GILT/IFI30 The Recombinant Human GILT/IFI30 Protein is derived from E. coli. The Recombinant Human GILT/IFI30 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
1mg/mL
20mM Tris-HCl buffer (pH 8.0) containing 0.1M NaCl, 10% glycerol, 1mM DTT
22.5 kDa
>90%, by SDS-PAGE
E. coli
IFI30
SDS-Page
10437
EC 1.8, Gamma-interferon-inducible protein IP-30, GILTgamma-interferon-inducible lysosomal thiol reductase, IFI-30, interferon, gamma-inducible protein 30, IP30interferon gamma-inducible protein 30 preproprotein, Legumaturain, MGC32056
0.25mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Human