Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Granzyme K Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$280.50 - $1004.00

Specifications

Source Baculovirus
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol GZMK
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25956150 View Documents Novus Biologicals™
NBP25956150UG
50μg Each for $280.50
Add to cart
 
NBP259561Y View Documents Novus Biologicals™
NBP259561250UG
250μg Each for $1,004.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 27-264 of Human Granzyme K The Recombinant Human Granzyme K Protein is derived from Baculovirus. The Recombinant Human Granzyme K Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Baculovirus
RUO
GZMK
3003
Recombinant Human Granzyme K Protein
Liquid. PBS (pH 7.4) containing 20% glycerol, 1mM DTT.
Unlabeled
0.25mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Fragmentin-3, granzyme 3, granzyme K, granzyme K (granzyme 3; tryptase II), granzyme K (serine protease, granzyme 3; tryptase II), Granzyme-3, NK-Tryp-2, NK-tryptase-2, PRSS, TRYP2granzyme-3, tryptase II
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
26.9 kDa
<1 EU per 1μg (determined by LAL method)

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit