Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Recombinant Human Growth Factors

Mediates the process of clonal expansion and maintenance of T-cell proliferation

Specifications

Cross Reactivity E. coli
Source Human Jurkat Cell Line
Type Recombinant Human Interleukin-2
For Use With (Application) Research use only
Stability 1 Year
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
22-157-347
View Documents
ZeptoMetrix
0801114
20000IU
Each for $101.70
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-389
View Documents
ZeptoMetrix
0802001
100μg
Each for $373.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

CELLKINES rhIL-2 originates from the IL-2 gene found in human Jurkat cell line which was cloned into E. coli. This product differs from native IL-2 in two ways. First, it is not glycosylated, and second, the protein differs at two amino acid positions (the first three N-terminal amino acids are absent, and CYS at position 125 is substituted by SER). CELLKINES rhIL-2 with a molecular weight of 15.4 kDa is formulated in 2% fetal bovine serum (heat inactivated) as a stabilizer and sterile filtered.

  • High specific activity and purity

Specifications

Specifications

E. coli
Recombinant Human Interleukin-2
1 Year
Human Jurkat Cell Line
Research use only
-20°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit