Learn More
R&D Systems™ Recombinant Human His6-Miro1/Rhot1 Protein, Carrier-free Available on GSA/VA Contract for Federal Government customers only.
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ SP490100
Catalog No. SP490100
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human His6-Miro1/Rhot1 Protein is derived from E. coli. The Recombinant Human His6-Miro1/Rhot1 Protein has been validated for the following applications: Bioactivity.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
The product is shipped with dry ice or equivalent.
Specifications
Specifications
Q8IXI2
Xmg/mL (XμM) in 50mM HEPES pH 8.0, 200mM NaCl, 10% (v/v) Glycerol, 2mM TCEP
50 kDa
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie Blue stain.
E. coli
RUO
Unconjugated
Yes
Bioactivity
55288
ARHT1, EC 3.6.5, EC 3.6.5.-, FLJ11040, FLJ12633, hMiro-1, MIRO-1mitochondrial Rho GTPase 1, mitochondrial Rho 1, rac-GTP binding protein-like protein, Rac-GTP-binding protein-like protein, Ras homolog gene family member T1, ras homolog gene family, member
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Measured in an anti-viral assay using HepG2 human hepatocellular carcinoma cells infected with encephalomyocarditis (EMC) virus. Sheppard, P. et al. (2003) Nat. Immunol. 4:63. The ED50 for this effect is 0.2-1.2 μg/mL.
Human