Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Histamine N-Methyltransferase/HNMT Isoform 1 Protein
SDP

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page, Bioactivity

$191.70 - $494.50

Specifications

Concentration 1 mg/ml
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris-Hcl (pH8.0) buffer containing 10% glycerol
Gene ID (Entrez) 3176
Molecular Weight (g/mol) 37.4 kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP26198020
SDP
View Documents
Novus Biologicals™
NBP26198020UG
20 μg
Each for $494.50
Add to Cart
 
NBP2619805
SDP
View Documents
Novus Biologicals™
NBP2619805UG
5 μg
Each for $191.70
Add to Cart
 
Description

Description

A Biologically active recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-292 of Human Histamine N-Methyltransferase/HNMT The Recombinant Human Histamine N-Methyltransferase/HNMT Isoform 1 Protein is derived from E. coli. The Recombinant Human Histamine N-Methyltransferase/HNMT Isoform 1 Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications

Specifications

1 mg/ml
20mM Tris-Hcl (pH8.0) buffer containing 10% glycerol
37.4 kDa
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
HNMT
Human
Bioactivity, SDS-PAGE
3176
E. coli
RUO
Histamine N-Methyltransferase/HNMT
>90%, by SDS-PAGE
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit