Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human IL-10 (aa 19-178) Protein, CF
Measured in a cell proliferation assay using MC/9-2 mouse mast cells. The ED50 for this effect is 0.15 to 0.75 ng/mL.
Supplier: R&D Systems™ 1064IL025/CF
This item is not returnable.
View return policy
Description
Measured in a cell proliferation assay using MC/9-2 mouse mast cells. Thompson-Snipes, L. et al. (1991) J. Exp. Med. 173:507. The ED50 for this effect is 0.075-0.750 ng/mL.Specifications
P22301 | |
3586 | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
E. coli-derived human IL-10 protein Ser19-Asn178, with an N-terminal Met | |
IL10 | |
Human | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Bioactivity | |
18.8 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. | |
Unconjugated | |
Reconstitute at 100 μg/mL in sterile PBS. | |
<0.10 EU per 1 μg of the protein by the LAL method. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction